Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNGA3
Entrez Gene ID
396144
Gene Name
cyclic nucleotide gated channel alpha 3
Gene Aliases
-
Chromosome Location
Chr.1: 133234443 - 133268781 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205221.1 NP_990552.1 7 - 8 806 102
XM_015277691.2 XP_015133177.1 5 - 6 936 102
GenBank mRNA X89598.1 - - 806 102

View Full Product Details