Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNNI2
Entrez Gene ID
396386
Gene Name
troponin I2, fast skeletal type
Gene Aliases
TNI
Chromosome Location
Chr.5: 14291354 - 14301164 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205417.1 NP_990748.1 4 - 5 219 68
GenBank mRNA U19926.1 - 4 - 5 219 68

View Full Product Details