Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSME3
Entrez Gene ID
420017
Gene Name
proteasome activator subunit 3
Gene Aliases
-
Chromosome Location
Chr.27: 7880094 - 7886800 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012550.2 NP_001012568.1 7 - 8 548 89
GenBank mRNA AJ719711.1 - 7 - 8 556 89
AJ851655.1 - 7 - 8 549 89

View Full Product Details