Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HNRNPH3
Entrez Gene ID
423686
Gene Name
heterogeneous nuclear ribonucleoprotein H3
Gene Aliases
HNRPH3
Chromosome Location
Chr.6: 11282248 - 11288755 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012592.1 NP_001012610.1 4 - 5 653 130
XM_015278681.2 XP_015134167.1 4 - 5 608 130
XM_015278683.2 XP_015134169.1 4 - 5 651 130
XM_025151554.1 XP_025007322.1 4 - 5 532 130
XM_025151556.1 XP_025007324.1 4 - 5 584 130
XM_025151562.1 XP_025007330.1 3 - 4 522 130
GenBank mRNA AJ851718.1 - 4 - 5 653 130

View Full Product Details