Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDUFAF5
Entrez Gene ID
416742
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 5
Gene Aliases
C20ORF7, C3H20orf7, bA526K24.2, dJ842G6.1
Chromosome Location
Chr.3: 12687574 - 12693226 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278083.1 NP_001265012.1 10 - 11 971 95
XM_025148612.1 XP_025004380.1 8 - 9 829 95
GenBank mRNA AJ851408.1 - 8 - 9 1123 95

View Full Product Details