Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NKRF
Entrez Gene ID
422370
Gene Name
NFKB repressing factor
Gene Aliases
-
Chromosome Location
Chr.4: 16533304 - 16541599 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012887.2 NP_001012905.2 1 - 2 517 71
GenBank mRNA AJ720987.1 - 1 - 2 517 71

View Full Product Details