Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KRT5
Entrez Gene ID
407779
Gene Name
keratin 5
Gene Aliases
-
Chromosome Location
Chr.33: 5260250 - 5267112 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001195.2 NP_001001195.1 2 - 3 882 82
GenBank mRNA AB734685.1 - 2 - 3 790 82
AB734686.1 - 2 - 3 790 82
AB734687.1 - 2 - 3 790 82
AY574983.1 - 2 - 3 882 82

View Full Product Details