Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTDSPL
Entrez Gene ID
408252
Gene Name
CTD small phosphatase like
Gene Aliases
NFI1, NIF
Chromosome Location
Chr.2: 4568816 - 4640232 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001316.1 NP_001001316.1 1 - 2 127 88
XM_015281052.2 XP_015136538.1 1 - 2 151 88
XM_025147078.1 XP_025002846.1 1 - 2 581 88
XM_025147079.1 XP_025002847.1 1 - 2 584 88
GenBank mRNA AF189773.1 - 1 - 2 127 88
AF189774.1 - 1 - 2 77 88
AF189775.1 - 1 - 2 131 88
AF189776.1 - 1 - 2 100 88

View Full Product Details