Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LYG2
Entrez Gene ID
395708
Gene Name
lysozyme g2
Gene Aliases
LYG1
Chromosome Location
Chr.1: 133828098 - 133831585 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001470.1 NP_001001470.1 2 - 3 191 115
XM_015277690.2 XP_015133176.1 2 - 3 185 115
GenBank mRNA X61002.1 - - 191 115
X61198.1 - - 191 115

View Full Product Details