Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYG2
Entrez Gene ID
395708
Gene Name
lysozyme g2
Gene Aliases
LYG1
Chromosome Location
Chr.1: 133828098 - 133831585 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001470.1 NP_001001470.1 5 - 6 635 64
XM_015277690.2 XP_015133176.1 5 - 6 629 64
GenBank mRNA X61002.1 - - 635 64
X61198.1 - - 635 64

View Full Product Details