Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LY86
Entrez Gene ID
420872
Gene Name
lymphocyte antigen 86
Gene Aliases
MD1
Chromosome Location
Chr.2: 64890313 - 64913904 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004399.1 NP_001004399.1 3 - 4 343 77
GenBank mRNA X60450.1 - - 343 77

View Full Product Details