Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDGFB
Entrez Gene ID
374128
Gene Name
platelet derived growth factor subunit B
Gene Aliases
-
Chromosome Location
Chr.1: 50720076 - 50737739 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204270.4 NP_989601.2 6 - 7 895 152
XM_015284373.2 XP_015139859.2 7 - 8 1747 152
XM_015284377.2 XP_015139863.2 7 - 8 1650 152
GenBank mRNA AB031025.1 - 6 - 7 856 152

View Full Product Details