Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD8BP
Entrez Gene ID
396175
Gene Name
CD8b molecule pseudogene
Gene Aliases
CD8B, CD8B1
Chromosome Location
Chr.4: 85919845 - 85924937 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205247.2 NP_990578.2 2 - 3 803 85
GenBank mRNA Y11475.1 - - 403 85
Z26484.1 - - 803 85

View Full Product Details