Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
YIPF4
Entrez Gene ID
421464
Gene Name
Yip1 domain family member 4
Gene Aliases
-
Chromosome Location
Chr.3: 31787547 - 31801055 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031058.1 NP_001026229.1 1 - 2 255 145
GenBank mRNA AJ720497.1 - 1 - 2 255 145

View Full Product Details