Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SASH3
Entrez Gene ID
422138
Gene Name
SAM and SH3 domain containing 3
Gene Aliases
CXorf9
Chromosome Location
Chr.4: 1633931 - 1638089 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031110.1 NP_001026281.1 7 - 8 1057 152
GenBank mRNA AJ720161.1 - 7 - 8 1057 152

View Full Product Details