Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD40
Entrez Gene ID
428678
Gene Name
ankyrin repeat domain 40
Gene Aliases
-
Chromosome Location
Chr.18: 10130720 - 10137479 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031531.1 NP_001026702.1 2 - 3 291 136
XM_015295417.2 XP_015150903.1 2 - 3 397 136
XM_015295418.2 XP_015150904.1 2 - 3 396 136
XM_015295419.2 XP_015150905.1 2 - 3 396 136
GenBank mRNA AJ720204.1 - 2 - 3 291 136

View Full Product Details