The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031605.1 | NP_001026776.1 | 3 - 4 | 380 | 142 |
GenBank mRNA | AJ719362.1 | - | 3 - 4 | 380 | 142 |