Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KAT7
Entrez Gene ID
425530
Gene Name
lysine acetyltransferase 7
Gene Aliases
MYST2
Chromosome Location
Chr.27: 5705539 - 5717026 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031341.1 NP_001026512.1 14 - 15 1938 111
XM_025143981.1 XP_024999749.1 13 - 14 1711 111
GenBank mRNA AJ720268.1 - 14 - 15 1938 111

View Full Product Details