Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPA
Entrez Gene ID
426329
Gene Name
protein phosphatase 2 phosphatase activator
Gene Aliases
PPP2R4
Chromosome Location
Chr.17: 5807399 - 5830881 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031371.1 NP_001026542.1 7 - 8 711 137
GenBank mRNA AJ719528.1 - 7 - 8 711 137

View Full Product Details