Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARSK
Entrez Gene ID
427116
Gene Name
arylsulfatase family member K
Gene Aliases
-
Chromosome Location
Chr.Z: 57617708 - 57628950 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031415.1 NP_001026586.1 5 - 6 911 104
GenBank mRNA AJ720194.1 - 5 - 6 911 104

View Full Product Details