Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COPS5
Entrez Gene ID
426579
Gene Name
COP9 signalosome subunit 5
Gene Aliases
-
Chromosome Location
Chr.2: 115315587 - 115326952 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039311.1 NP_001034400.1 3 - 4 649 141
GenBank mRNA AJ719811.1 - 3 - 4 649 141

View Full Product Details