Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RDM1
Entrez Gene ID
395228
Gene Name
RAD52 motif containing 1
Gene Aliases
RAD52B
Chromosome Location
Chr.27: 4030005 - 4034837 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204546.2 NP_989877.2 2 - 3 279 68
XM_015299306.2 XP_015154792.1 2 - 3 278 68
XM_015299307.2 XP_015154793.1 2 - 3 287 68
GenBank mRNA AB080727.1 - 2 - 3 279 68
BX930281.2 - 1 - 2 228 68
BX932627.1 - 2 - 3 276 68

View Full Product Details