Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD1C
Entrez Gene ID
425801
Gene Name
CD1c molecule
Gene Aliases
CD1.1, CD1.2, CD1A1, CD1D
Chromosome Location
Chr.16: 2665728 - 2668557 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017412.1 NP_001017412.1 1 - 2 106 99
GenBank mRNA AB204801.1 - 1 - 2 106 99
AY874074.1 - 1 - 2 61 99
AY874075.1 - 1 - 2 61 99
AY874077.1 - 1 - 2 61 99

View Full Product Details