Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EEF1A2
Entrez Gene ID
419244
Gene Name
eukaryotic translation elongation factor 1 alpha 2
Gene Aliases
-
Chromosome Location
Chr.20: 9001255 - 9014412 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032398.4 NP_001027570.3 5 - 6 1574 76
XM_015296389.2 XP_015151875.2 5 - 6 885 76
XM_025142223.1 XP_024997991.1 5 - 6 952 76
GenBank mRNA BX934090.2 - 3 - 4 76
DR427690.1 - 2 - 3 171 76

View Full Product Details