Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL10RA
Entrez Gene ID
419771
Gene Name
interleukin 10 receptor subunit alpha
Gene Aliases
IL10R1
Chromosome Location
Chr.24: 5431062 - 5434652 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039597.1 NP_001034686.1 2 - 3 213 85
GenBank mRNA AM049243.1 - 2 - 3 213 85
AM049244.1 - 2 - 3 213 85

View Full Product Details