Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCR2
Entrez Gene ID
420696
Gene Name
C-C motif chemokine receptor 2
Gene Aliases
CCR5-L
Chromosome Location
Chr.2: 42632311 - 42640006 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045835.1 NP_001039300.1 1 - 1 852 90
XM_015281503.2 XP_015136989.1 3 - 3 1157 90

View Full Product Details