Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP51A1
Entrez Gene ID
420548
Gene Name
cytochrome P450 family 51 subfamily A member 1
Gene Aliases
CYP51
Chromosome Location
Chr.2: 22493945 - 22511501 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048077.1 NP_001041542.1 6 - 7 673 132
XM_015281391.2 XP_015136877.1 4 - 5 624 132
XM_015281392.2 XP_015136878.1 8 - 9 2945 132
GenBank mRNA AB243875.1 - 6 - 7 673 132
AB243876.1 - 4 - 5 486 132

View Full Product Details