Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HMGN5
Entrez Gene ID
422278
Gene Name
high mobility group nucleosome binding domain 5
Gene Aliases
-
Chromosome Location
Chr.4: 9382554 - 9391441 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015278521.2 XP_015134007.1 3 - 4 250 81
XM_015278522.2 XP_015134008.1 2 - 3 327 81
GenBank mRNA EF422210.1 - 2 - 3 288 81

View Full Product Details