Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BORA
Entrez Gene ID
769298
Gene Name
bora, aurora kinase A activator
Gene Aliases
C13orf34, C1H13orf34
Chromosome Location
Chr.1: 157621738 - 157641309 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079741.2 NP_001073209.1 10 - 11 1593 120
XM_015275799.2 XP_015131285.1 12 - 13 1958 120
XM_015275801.2 XP_015131287.1 11 - 12 1770 120
XM_015275815.2 XP_015131301.1 12 - 13 1825 120
XM_025151504.1 XP_025007272.1 12 - 13 2009 120
XM_025151507.1 XP_025007275.1 9 - 10 1596 120
GenBank mRNA AJ851579.1 - 10 - 11 1594 120

View Full Product Details