Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCKAR
Entrez Gene ID
422801
Gene Name
cholecystokinin A receptor
Gene Aliases
-
Chromosome Location
Chr.4: 73203371 - 73210217 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081501.1 NP_001074970.1 3 - 4 998 98
GenBank mRNA AB214534.1 - 3 - 4 998 98

View Full Product Details