Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EDA2R
Entrez Gene ID
422166
Gene Name
ectodysplasin A2 receptor
Gene Aliases
-
Chromosome Location
Chr.4: 382939 - 391887 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083360.1 NP_001076829.1 5 - 6 592 69
GenBank mRNA DQ360501.1 - 5 - 6 592 69

View Full Product Details