Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THOC7
Entrez Gene ID
416077
Gene Name
THO complex 7
Gene Aliases
-
Chromosome Location
Chr.12: 13920134 - 13927881 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098238.3 NP_001091708.3 3 - 4 286 76

View Full Product Details