Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RGS17
Entrez Gene ID
395645
Gene Name
regulator of G-protein signaling 17
Gene Aliases
-
Chromosome Location
Chr.3: 49724363 - 49791838 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204841.2 NP_990172.1 2 - 3 304 74
GenBank mRNA AF151968.2 - 2 - 3 304 74

View Full Product Details