Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANP32E
Entrez Gene ID
426109
Gene Name
acidic nuclear phosphoprotein 32 family member E
Gene Aliases
-
Chromosome Location
Chr.25: 640387 - 652544 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006564.2 NP_001006564.2 3 - 4 470 152
XM_015298326.2 XP_015153812.1 3 - 4 541 152
GenBank mRNA AJ720498.1 - 3 - 4 506 152
AJ851397.1 - 3 - 4 470 152

View Full Product Details