The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_204162.1 | NP_989493.1 | 1 - 2 | 395 | 82 |
GenBank mRNA | AF092538.1 | - | 1 - 2 | 395 | 82 |