Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TWSG1
Entrez Gene ID
374024
Gene Name
twisted gastrulation BMP signaling modulator 1
Gene Aliases
-
Chromosome Location
Chr.2: 98562426 - 98580536 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204198.1 NP_989529.1 4 - 5 544 71
XM_025146996.1 XP_025002764.1 4 - 5 565 71
GenBank mRNA AF255731.1 - 4 - 5 544 71

View Full Product Details