Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CBX1
Entrez Gene ID
374237
Gene Name
chromobox 1
Gene Aliases
-
Chromosome Location
Chr.27: 6468583 - 6476595 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204332.1 NP_989663.1 2 - 3 281 85
XM_015299477.2 XP_015154963.1 3 - 4 422 85
XM_015299478.2 XP_015154964.1 2 - 3 323 85
GenBank mRNA AB005617.1 - 2 - 3 281 85

View Full Product Details