Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCX
Entrez Gene ID
374242
Gene Name
doublecortin
Gene Aliases
18C15.5
Chromosome Location
Chr.4: 13185758 - 13314342 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001305674.2 NP_001292603.1 5 - 6 1130 64
NM_204335.3 NP_989666.1 5 - 6 1130 64
XM_015278178.2 XP_015133664.1 8 - 9 3611 64
XM_015278179.2 XP_015133665.1 8 - 9 3610 64
GenBank mRNA AF330009.1 - 5 - 6 1016 64
CR406311.1 - 2 - 3 190 64

View Full Product Details