Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
DDAH1
Entrez Gene ID
378898
Gene Name
dimethylarginine dimethylaminohydrolase 1
Gene Aliases
-
Chromosome Location
Chr.8: 16625972 - 16676035 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204382.1 NP_989713.1 4 - 5 642 65
XM_025152732.1 XP_025008500.1 4 - 5 313 65
GenBank mRNA AB095027.2 - 4 - 5 642 65

View Full Product Details