Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLA
Entrez Gene ID
378908
Gene Name
Src like adaptor
Gene Aliases
src-like-adapter
Chromosome Location
Chr.2: 141829289 - 141850007 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204391.1 NP_989722.1 1 - 2 68 69
XM_015283077.2 XP_015138563.1 2 - 3 270 69
XM_015283079.2 XP_015138565.1 2 - 3 255 69
GenBank mRNA AJ721107.1 - 1 - 2 250 69
AY278230.1 - 1 - 2 68 69

View Full Product Details