Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBX18
Entrez Gene ID
395104
Gene Name
T-box transcription factor 18
Gene Aliases
-
Chromosome Location
Chr.3: 77240681 - 77263108 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204453.4 NP_989784.2 4 - 5 712 82
GenBank mRNA AY173127.2 - 4 - 5 712 82
AY550994.1 - 1 - 2 122 82

View Full Product Details