Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HNRNPH1
Entrez Gene ID
395157
Gene Name
heterogeneous nuclear ribonucleoprotein H1
Gene Aliases
HNRNPH2, HNRPH1
Chromosome Location
Chr.13: 14281474 - 14298029 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204496.1 NP_989827.1 3 - 4 466 76
XM_015293722.2 XP_015149208.2 2 - 3 407 76
XM_015293723.2 XP_015149209.2 4 - 5 455 76
XM_015293725.2 XP_015149211.2 4 - 5 558 76
XM_015293726.2 XP_015149212.2 3 - 4 395 76
XM_015293728.2 XP_015149214.2 3 - 4 327 76
XM_015293730.2 XP_015149216.2 3 - 4 466 76
XM_025154783.1 XP_025010551.1 3 - 4 466 76
XM_025154784.1 XP_025010552.1 4 - 5 435 76
XM_025154785.1 XP_025010553.1 3 - 4 466 76
XM_025154786.1 XP_025010554.1 3 - 4 255 76
XM_025154787.1 XP_025010555.1 3 - 4 469 76
XM_025154788.1 XP_025010556.1 3 - 4 466 76
XM_025154789.1 XP_025010557.1 3 - 4 496 76
XM_025154790.1 XP_025010558.1 4 - 5 392 76
XM_025154791.1 XP_025010559.1 2 - 3 407 76
XM_025154792.1 XP_025010560.1 3 - 4 374 76
XM_025154793.1 XP_025010561.1 2 - 3 424 76
XM_025154794.1 XP_025010562.1 2 - 3 407 76
XM_025154795.1 XP_025010563.1 3 - 4 466 76
XM_025154796.1 XP_025010564.1 4 - 5 384 76
XM_025154797.1 XP_025010565.1 4 - 5 558 76
GenBank mRNA AJ719489.1 - 2 - 3 403 76
AJ851662.1 - 3 - 4 367 76
AY278548.1 - 3 - 4 466 76

View Full Product Details