Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HNRNPH1
Entrez Gene ID
395157
Gene Name
heterogeneous nuclear ribonucleoprotein H1
Gene Aliases
HNRNPH2, HNRPH1
Chromosome Location
Chr.13: 14281474 - 14298029 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204496.1 NP_989827.1 7 - 8 1078 83
XM_015293722.2 XP_015149208.2 6 - 7 1019 83
XM_015293723.2 XP_015149209.2 8 - 9 1067 83
XM_015293725.2 XP_015149211.2 8 - 9 1170 83
XM_015293726.2 XP_015149212.2 7 - 8 1007 83
XM_015293728.2 XP_015149214.2 7 - 8 939 83
XM_025154783.1 XP_025010551.1 7 - 8 1078 83
XM_025154784.1 XP_025010552.1 8 - 9 1047 83
XM_025154785.1 XP_025010553.1 7 - 8 1078 83
XM_025154786.1 XP_025010554.1 7 - 8 867 83
XM_025154787.1 XP_025010555.1 7 - 8 1081 83
XM_025154788.1 XP_025010556.1 7 - 8 1078 83
XM_025154789.1 XP_025010557.1 7 - 8 1108 83
XM_025154790.1 XP_025010558.1 8 - 9 1004 83
XM_025154791.1 XP_025010559.1 6 - 7 1019 83
XM_025154792.1 XP_025010560.1 7 - 8 986 83
XM_025154793.1 XP_025010561.1 6 - 7 1036 83
XM_025154794.1 XP_025010562.1 6 - 7 1019 83
XM_025154797.1 XP_025010565.1 8 - 9 1170 83
GenBank mRNA AJ719489.1 - 6 - 7 1015 83
AJ851662.1 - 7 - 8 979 83
AY278548.1 - 7 - 8 1078 83

View Full Product Details