Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DHRS11
Entrez Gene ID
395172
Gene Name
dehydrogenase/reductase 11
Gene Aliases
-
Chromosome Location
Chr.19: 8314191 - 8376854 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204507.1 NP_989838.1 6 - 7 735 85
XM_015296153.2 XP_015151639.1 5 - 6 924 85
GenBank mRNA AF373778.1 - 6 - 7 735 85
BX933105.1 - 6 - 7 614 85
BX936181.1 - 4 - 5 388 85

View Full Product Details