Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MATN3
Entrez Gene ID
395954
Gene Name
matrilin 3
Gene Aliases
matrilin-3
Chromosome Location
Chr.3: 101394465 - 101409742 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205072.1 NP_990403.1 6 - 7 1224 71
GenBank mRNA AJ000055.1 - 6 - 7 1224 71
CR389007.1 - 2 - 3 196 71
CR390328.1 - 2 - 3 190 71

View Full Product Details