Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NEUROD4
Entrez Gene ID
395959
Gene Name
neuronal differentiation 4
Gene Aliases
-
Chromosome Location
Chr.33: 5981077 - 5986673 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205076.1 NP_990407.1 1 - 1 1048 93
XM_015300228.2 XP_015155714.1 2 - 2 1719 93
GenBank mRNA Y09597.1 - - 1048 93

View Full Product Details