Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TNNC1
Entrez Gene ID
396032
Gene Name
troponin C1, slow skeletal and cardiac type
Gene Aliases
PCTNC1
Chromosome Location
Chr.12: 975844 - 981282 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205133.1 NP_990464.1 4 - 5 331 121
GenBank mRNA D13037.1 - 4 - 5 331 121
M16024.1 - 4 - 5 332 121

View Full Product Details