Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYL3
Entrez Gene ID
396067
Gene Name
myosin, light chain 3, alkali; ventricular, skeletal, slow
Gene Aliases
MLC, MLC1c/1s
Chromosome Location
Chr.2: 3512669 - 3547290 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205159.1 NP_990490.1 3 - 4 364 79
XM_015281049.2 XP_015136535.1 3 - 4 407 79
XM_015281050.2 XP_015136536.1 3 - 4 400 79
GenBank mRNA X13861.1 - - 304 79
X13862.1 - - 364 79

View Full Product Details