Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCL
Entrez Gene ID
396201
Gene Name
nucleolin
Gene Aliases
-
Chromosome Location
Chr.9: 15430541 - 15437975 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205265.1 NP_990596.1 8 - 9 1201 112
GenBank mRNA X17199.1 - - 1201 112

View Full Product Details