Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOPNL
Entrez Gene ID
416598
Gene Name
FGFR1OP N-terminal like
Gene Aliases
C14H16orf63, C16orf63, CEP20
Chromosome Location
Chr.14: 8032225 - 8037255 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006170.1 NP_001006170.1 4 - 5 463 67
XM_015294217.2 XP_015149703.1 4 - 5 504 67
GenBank mRNA AJ720610.1 - 4 - 5 463 67

View Full Product Details